N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF672.5 008406-5G 008406-25G 008406-1G 008406-10G
CHMPCFF672.5EA 0 GBP
CHMPCFF672.5 FLUO008406-5G FLUO008406-25G FLUO008406-1G FLUO008406-10G
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine
Formula: C₁₂H₁₃F₃N₂O₂
MDL Number: MFCD00052908
CAS Number: 1692-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR