5-Bromo-4-butylpyrimidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6072-5G HC-6072-1G
COBBHC-6072-5GEA 0 GBP
COBBHC-6072-5G COBBHC-6072-1G
5-Bromo-4-butylpyrimidine
5-Bromo-4-butylpyrimidine
Formula: C₈H₁₁BrN₂
MDL Number: MFCD09800931
CAS Number: 4595-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR