1-Bromo-4-methoxynaphthalene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0651-5G OT-0651-1G
COBBOT-0651-5GEA 0 GBP
COBBOT-0651-5G COBBOT-0651-1G
1-Bromo-4-methoxynaphthalene
1-Bromo-4-methoxynaphthalene
Formula: C₁₁H₉BrO
MW: 237.0926 g/mol
MDL Number: MFCD00155144
CAS Number: 5467-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR