1-Bromo-4-(trans-4-ethylcyclohexyl)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7496-25G OS-7496-1G OS-7496-5G
COBBOS-7496-25GEA 0 GBP
COBBOS-7496-25G COBBOS-7496-1G COBBOS-7496-5G
1-Bromo-4-(trans-4-ethylcyclohexyl)benzene
1-Bromo-4-(trans-4-ethylcyclohexyl)benzene
Formula: C₁₄H₁₉Br
MDL Number: MFCD08275628
CAS Number: 91538-82-8
EINECS: 422-030-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR