4-Chloro-o-anisidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1172-5G AN-1172-1G
COBBAN-1172-5GEA 0 GBP
COBBAN-1172-5G COBBAN-1172-1G
4-Chloro-o-anisidine
4-Chloro-o-anisidine
Formula: C₇H₈ClNO
MDL Number: MFCD06801386
CAS Number: 93-50-5
EINECS: 202-251-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR