5-Bromo-2-(2-methoxyethoxy)anisole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1162-5G OT-1162-1G
COBBOT-1162-5GEA 0 GBP
COBBOT-1162-5G COBBOT-1162-1G
5-Bromo-2-(2-methoxyethoxy)anisole
5-Bromo-2-(2-methoxyethoxy)anisole
Formula: C₁₀H₁₃BrO₃
MDL Number: MFCD15143589
CAS Number: 1092563-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR