6-Chloro-2-fluoro-3-methylanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-3-fluoro-2-methoxy-4-methylbenzene

005641-1G 005641-5G
FLUO005641-1GEA 0 GBP
FLUO005641-1G FLUO005641-5G
6-Chloro-2-fluoro-3-methylanisole
6-Chloro-2-fluoro-3-methylanisole
Formula: C₈H₈ClFO
MW: 174.60 g/mol
MDL Number: MFCD01631377
CAS Number: 261762-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR