4,5-Dimethyl-2-ethylthiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009626-1G 009626-5G
FLUO009626-1GEA 0 GBP
FLUO009626-1G FLUO009626-5G
4,5-Dimethyl-2-ethylthiazole
4,5-Dimethyl-2-ethylthiazole
Formula: C₇H₁₁NS
MDL Number: MFCD00053125
CAS Number: 873-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR