4,5-Dimethyl-2-isobutylthiazole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009627-1G
009627-5G
FLUO009627-1GEA
0
GBP
FLUO009627-1G
FLUO009627-5G
4,5-Dimethyl-2-isobutylthiazole
4,5-Dimethyl-2-isobutylthiazole
Formula:
C₉H₁₅NS MW: 169.28 g/mol |
MDL Number:
MFCD00053136 CAS Number: 53498-32-1 EINECS: 258-586-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...