N-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF671.5
008405-5G
008405-1G
CHMPCFF671.5EA
0
GBP
CHMPCFF671.5
FLUO008405-5G
FLUO008405-1G
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
Formula:
C₁₁H₁₂F₃N₃O₂ |
MDL Number:
MFCD00052613 CAS Number: 58315-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...