Tetrafluoroisophthalic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Perfluoroisophthalic acid, 2,4,5,6-Tetrafluoroisophthalic acid
M-Clarity™ quality level = MQ100
337668-1G
337668-5G
SIAL337668-1GEA
206
GBP
SIAL337668-1G
SIAL337668-5G
Tetrafluoroisophthalic acid, Sigma-Aldrich®
Tetrafluoroisophthalic acid
Formula:
C₈H₂F₄O₄ MW: 238.0927 g/mol |
MDL Number:
MFCD00042379 CAS Number: 1551-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...