Ethyl-α-bromophenylacetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl alpha-bromophenylacetate

M-Clarity™ quality level = MQ100

554065-100G 554065-25G
SIAL554065-100GEA 758 GBP
SIAL554065-100G SIAL554065-25G
Ethyl-α-bromophenylacetate, Sigma-Aldrich®
Ethyl α-bromophenylacetate
Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD00013536
CAS Number: 2882-19-1
EINECS: 220-735-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR