2-Methyl-3-(trifluoromethyl)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009858-1G 009858-5G
FLUO009858-1GEA 0 GBP
FLUO009858-1G FLUO009858-5G
2-Methyl-3-(trifluoromethyl)benzoic acid
2-Methyl-3-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
MDL Number: MFCD01631588
CAS Number: 62089-35-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR