trans-1,3-Pentadiene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

111805-1G 111805-5G
SIAL111805-1GEA 149 GBP
SIAL111805-1G SIAL111805-5G
trans-1,3-Pentadiene, Sigma-Aldrich®
trans-1,3-Pentadiene
Formula: C₅H₈
MDL Number: MFCD00009295
CAS Number: 2004-70-8
EINECS: 217-909-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR