6-Fluoro-o-toluidine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
012747-1G
012747-5G
FLUO012747-1GEA
0
GBP
FLUO012747-1G
FLUO012747-5G
6-Fluoro-o-toluidine
6-Fluoro-o-toluidine
Formula:
C₇H₈FN |
MDL Number:
MFCD06658252 CAS Number: 443-89-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...