4-(Difluoromethoxy)benzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004833-5G
004833-1G
FLUO004833-5GEA
0
GBP
FLUO004833-5G
FLUO004833-1G
4-(Difluoromethoxy)benzonitrile
4-(Difluoromethoxy)benzonitrile
Formula:
C₈H₅F₂NO |
MDL Number:
MFCD00085006 CAS Number: 90446-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...