Chlorobis(4-methylphenyl)phosphine 95%, rods
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H25889.06
H25889.03
H25889.06EA
227
GBP
H25889.06
H25889.03
Chlorobis(4-methylphenyl)phosphine 95%, rods
Chlorobis(4-methylphenyl)phosphine
Purity: 95%
CAS No.: 1019-71-2
Molecular Formula: C14H14ClP
Formula Weight: 248.69
Boiling Point: 180-184°/10mm
UN No.: 1760
MDL No.: MFCD01630844
Formula:
C₁₄H₁₄ClP Boiling Pt: 180…184 °C (10 mmHg) Density: 1.159 Storage Temperature: Ambient |
MDL Number:
MFCD01630844 CAS Number: 1019-71-2 UN: 1760 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...