Bromomethyl methyl ether

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005432-25G 005432-100G
FLUO005432-25GEA 0 GBP
FLUO005432-25G FLUO005432-100G
Bromomethyl methyl ether
Bromomethyl methyl ether
Formula: C₂H₅BrO
MDL Number: MFCD00000171
CAS Number: 13057-17-5
UN: 1992
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR