N-[2-Nitro-4-(trifluoromethyl)phenyl]morpholine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF665.5 008404-5G 008404-25G
CHMPCFF665.5EA 0 GBP
CHMPCFF665.5 FLUO008404-5G FLUO008404-25G
N-[2-Nitro-4-(trifluoromethyl)phenyl]morpholine
N-[2-Nitro-4-(trifluoromethyl)phenyl]morpholine
Formula: C₁₁H₁₁F₃N₂O₃
MDL Number: MFCD00052375
CAS Number: 62054-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR