2,4-Diamino-6-nitroquinazoline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023271-1G 023271-5G
FLUO023271-1GEA 0 GBP
FLUO023271-1G FLUO023271-5G
2,4-Diamino-6-nitroquinazoline
2,4-Diamino-6-nitroquinazoline
Formula: C₈H₇N₅O₂
MW: 205.18 g/mol
MDL Number: MFCD00023910
CAS Number: 7154-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR