2,4-Dichloro-6-methoxyquinazoline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS2578G1
AS2578G5
ACTIAS2578G1EA
58
GBP
ACTIAS2578G1
ACTIAS2578G5
2,4-Dichloro-6-methoxyquinazoline
2,4-Dichloro-6-methoxyquinazoline
Formula:
C₉H₆Cl₂N₂O MW: 229.0627 g/mol |
MDL Number:
MFCD09954880 CAS Number: 105763-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...