2-Chloroquinazoline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS2470G1
AS2470G5
ACTIAS2470G1EA
57
GBP
ACTIAS2470G1
ACTIAS2470G5
2-Chloroquinazoline
2-Chloroquinazoline
Formula:
C₈H₅ClN₂ |
MDL Number:
MFCD09959715 CAS Number: 6141-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...