2-Chloro-3-nitro-5-thiophenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF258.2.5 CFF258.10 CFF258.25 005880-5G 005880-1G
CHMPCFF258.2.5EA 0 GBP
CHMPCFF258.2.5 CHMPCFF258.10 CHMPCFF258.25 FLUO005880-5G FLUO005880-1G
2-Chloro-3-nitro-5-thiophenesulphonyl chloride
2-Chloro-3-nitro-5-thiophenesulphonyl chloride
Formula: C₄HCl₂NO₄S₂
MDL Number: MFCD00067978
CAS Number: 58457-24-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR