2-Chloro-3-methylpyridine-N-oxide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7972-1G PY-7972-5G PY-7972-250M
COBBPY-7972-1GEA 0 GBP
COBBPY-7972-1G COBBPY-7972-5G COBBPY-7972-250M
2-Chloro-3-methylpyridine-N-oxide
2-Chloro-3-methylpyridine N-oxide
Formula: C₆H₆ClNO
MDL Number: MFCD14582951
CAS Number: 91668-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR