Nickel(II) tetrafluoroborate hexahydrate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006876-50G 006876-250G 006876-1KG
FLUO006876-50GEA 0 GBP
FLUO006876-50G FLUO006876-250G FLUO006876-1KG
Nickel(II) tetrafluoroborate hexahydrate
Nickel (II) tetrafluoroborate hexahydrate
Formula: B₂F₈Ni·6H₂O
MDL Number: MFCD00150259
CAS Number: 15684-36-3
EINECS: 238-753-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR