Cyclopropanesulphonamide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

674060-1G 674060-5G
SIAL674060-1GEA 98.5 GBP
SIAL674060-1G SIAL674060-5G
Cyclopropanesulphonamide, Sigma-Aldrich®
Cyclopropanesulphonamide
Formula: C₃H₇NO₂S
MW: 121.15 g/mol
MDL Number: MFCD08705286
CAS Number: 154350-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR