Cyclopropylboronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

M-Clarity™ quality level = MQ100

659851-1G 659851-5G
SIAL659851-1GEA 38.8 GBP
SIAL659851-1G SIAL659851-5G
Cyclopropylboronic acid pinacol ester, Sigma-Aldrich®
Cyclopropylboronic acid pinacol ester
Formula: C₉H₁₇BO₂
MDL Number: MFCD05663847
CAS Number: 126689-01-8
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR