Cyclopropylboronic acid pinacol ester
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
021608-5G
021608-1G
FLUO021608-5GEA
0
GBP
FLUO021608-5G
FLUO021608-1G
Cyclopropylboronic acid pinacol ester
Cyclopropylboronic acid pinacol ester
Formula:
C₉H₁₇BO₂ |
MDL Number:
MFCD05663847 CAS Number: 126689-01-8 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...