2-(Benzyloxy)-4-bromoanisole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0681-5G
OT-0681-1G
COBBOT-0681-5GEA
0
GBP
COBBOT-0681-5G
COBBOT-0681-1G
2-(Benzyloxy)-4-bromoanisole
2-(Benzyloxy)-4-bromoanisole
Formula:
C₁₄H₁₃BrO₂ |
MDL Number:
MFCD09836381 CAS Number: 78504-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...