4-(Benzyloxy)-3-bromo-5-fluoroaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1561-5G AN-1561-1G
COBBAN-1561-5GEA 0 GBP
COBBAN-1561-5G COBBAN-1561-1G
4-(Benzyloxy)-3-bromo-5-fluoroaniline
4-(Benzyloxy)-3-bromo-5-fluoroaniline
Formula: C₁₃H₁₁BrFNO
MDL Number: MFCD15143448
CAS Number: 1215206-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR