3,5-Bis(trifluoromethyl)phenylhydrazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006035-5G 006035-1G 006035-25G
FLUO006035-5GEA 0 GBP
FLUO006035-5G FLUO006035-1G FLUO006035-25G
3,5-Bis(trifluoromethyl)phenylhydrazine
3,5-Bis(trifluoromethyl)phenylhydrazine
Formula: C₈H₆F₆N₂
MW: 244.14 g/mol
MDL Number: MFCD00051606
CAS Number: 886-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR