2-(Methoxycarbonyl)-5-(trifluoromethyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-(Methoxycarbonyl)-5-(trifluoromethyl)phenyl)boronic acid

BB-5422-5G BB-5422-1G BB-5422-250M
COBBBB-5422-5GEA 0 GBP
COBBBB-5422-5G COBBBB-5422-1G COBBBB-5422-250M
2-(Methoxycarbonyl)-5-(trifluoromethyl)phenylboronic acid
2-(Methoxycarbonyl)-5-(trifluoromethyl)phenylboronic acid
Formula: C₉H₈BF₃O₄
MDL Number: MFCD12025964
CAS Number: 1217500-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR