3,5-Dimethyl-4-methoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Methoxy-3,5-dimethylphenylboronic acid, 4-Methoxy-3,5-dimethylbenzeneboronic acid

BB-2155-5G BB-2155-25G
COBBBB-2155-5GEA 0 GBP
COBBBB-2155-5G COBBBB-2155-25G
3,5-Dimethyl-4-methoxyphenylboronic acid
3,5-Dimethyl-4-methoxyphenylboronic acid
Formula: C₉H₁₃BO₃
MDL Number: MFCD01114648
CAS Number: 301699-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR