3,5-Dimethyl-4-isopropoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8116-5G BB-8116-1G BB-8116-25G
COBBBB-8116-5GEA 0 GBP
COBBBB-8116-5G COBBBB-8116-1G COBBBB-8116-25G
3,5-Dimethyl-4-isopropoxyphenylboronic acid
3,5-Dimethyl-4-isopropoxyphenylboronic acid
Formula: C₁₁H₁₇BO₃
MDL Number: MFCD06798063
CAS Number: 849062-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR