Potassium-4-(benzyloxy)-2-methylphenyltrifluoroborate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TB-3207-1G
TB-3207-5G
COBBTB-3207-1GEA
0
GBP
COBBTB-3207-1G
COBBTB-3207-5G
Potassium-4-(benzyloxy)-2-methylphenyltrifluoroborate
Potassium 4-(benzyloxy)-2-methylphenyltrifluoroborate
Formula:
C₁₄H₁₃BF₃KO |
MDL Number:
MFCD04115740 CAS Number: 850623-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...