4-Hydroxy-3-(trifluoromethyl)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013033-5G 013033-1G
FLUO013033-5GEA 0 GBP
FLUO013033-5G FLUO013033-1G
4-Hydroxy-3-(trifluoromethyl)benzoic acid
4-Hydroxy-3-(trifluoromethyl)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
MDL Number: MFCD01091008
CAS Number: 220239-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR