4-Hydroxy-2-(trifluoromethyl)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013956-1G 013956-5G
FLUO013956-1GEA 0 GBP
FLUO013956-1G FLUO013956-5G
4-Hydroxy-2-(trifluoromethyl)benzoic acid
4-Hydroxy-2-(trifluoromethyl)benzoic acid
Formula: C₈H₅F₃O₃
MDL Number: MFCD01091007
CAS Number: 320-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR