3-(Trifluoromethyl)benzoic acid hydrazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Trifluoromethyl)benzhydrazide, 3-(Trifluoromethyl)benzohydrazide

CFF689.5 CFF689.10 008430-1G 008430-5G
CHMPCFF689.5EA 0 GBP
CHMPCFF689.5 CHMPCFF689.10 FLUO008430-1G FLUO008430-5G
3-(Trifluoromethyl)benzoic acid hydrazide
3-(Trifluoromethyl)benzoic acid hydrazide
Formula: C₈H₇F₃N₂O
MW: 204.1492 g/mol
MDL Number: MFCD00221476
CAS Number: 22227-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR