4-(Trifluoroacetyl)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(2,2,2-trifluoroacetyl)benzoic acid

007472-1G 007472-5G
FLUO007472-1GEA 0 GBP
FLUO007472-1G FLUO007472-5G
4-(Trifluoroacetyl)benzoic acid
4-(Trifluoroacetyl)benzoic acid
Formula: C₉H₅F₃O₃
MW: 218.13 g/mol
MDL Number: MFCD00052340
CAS Number: 58808-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR