Methyl-5-acetylsalicylate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 5-acetyl-2-hydroxybenzoate

M-Clarity™ quality level = MQ200

390208-25G 390208-5G
SIAL390208-25GEA 74.8 GBP
SIAL390208-25G SIAL390208-5G
Methyl-5-acetylsalicylate, Sigma-Aldrich®
Methyl 5-acetylsalicylate
Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
MDL Number: MFCD00040924
CAS Number: 16475-90-4
EINECS: 240-532-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR