2-Chloro-4-methylphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Chloro-4-methylbenzeneboronic acid
BB-2194-5G
BB-2194-1G
BB-2194-25G
COBBBB-2194-5GEA
0
GBP
COBBBB-2194-5G
COBBBB-2194-1G
COBBBB-2194-25G
2-Chloro-4-methylphenylboronic acid
2-Chloro-4-methylphenylboronic acid
Formula:
C₇H₈BClO₂ |
MDL Number:
MFCD03411936 CAS Number: 145349-62-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...