4-Bromo-3-methylthioanisole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0561-5G
OT-0561-1G
COBBOT-0561-5GEA
0
GBP
COBBOT-0561-5G
COBBOT-0561-1G
4-Bromo-3-methylthioanisole
4-Bromo-3-methylthioanisole
Formula:
C₈H₉BrS |
MDL Number:
MFCD09801036 CAS Number: 90532-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...