Potassium-2-(hydroxymethyl)phenyltrifluoroborate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TB-3245-1G
TB-3245-25G
TB-3245-5G
COBBTB-3245-1GEA
0
GBP
COBBTB-3245-1G
COBBTB-3245-25G
COBBTB-3245-5G
Potassium-2-(hydroxymethyl)phenyltrifluoroborate
Potassium 2-(hydroxymethyl)phenyltrifluoroborate
Formula:
C₇H₇BF₃KO |
MDL Number:
MFCD04115769 CAS Number: 850623-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...