4-(Hydroxymethyl)phenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

016259-1G 016259-25G 016259-5G
FLUO016259-1GEA 0 GBP
FLUO016259-1G FLUO016259-25G FLUO016259-5G
4-(Hydroxymethyl)phenylboronic acid
4-(Hydroxymethyl)phenylboronic acid
Formula: C₇H₉BO₃
MDL Number: MFCD00792672
CAS Number: 59016-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR