4-Methoxycarbonyl-2-nitrophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Methoxycarbonyl)-2-nitrophenylboronic acid, 4-(Methoxycarbonyl)-2-nitrobenzeneboronic acid, 4-Methoxycarbonyl-2-nitrobenzeneboronic acid

BB-2461-5G BB-2461-25G BB-2461-1G
COBBBB-2461-5GEA 0 GBP
COBBBB-2461-5G COBBBB-2461-25G COBBBB-2461-1G
4-Methoxycarbonyl-2-nitrophenylboronic acid
4-Methoxycarbonyl-2-nitrophenylboronic acid
Formula: C₈H₈BNO₆
MW: 224.96 g/mol
MDL Number: MFCD01632202
CAS Number: 85107-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR