2-Amino-4-(4-fluorophenyl)thiazole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
010123-1G
010123-10G
FLUO010123-1GEA
0
GBP
FLUO010123-1G
FLUO010123-10G
2-Amino-4-(4-fluorophenyl)thiazole
2-Amino-4-(4-fluorophenyl)thiazole
Formula:
C₉H₇FN₂S MW: 194.2287 g/mol |
MDL Number:
MFCD01086598 CAS Number: 77815-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...