2-Amino-4-(4-fluorophenyl)thiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2045-5G HC-2045-1G
COBBHC-2045-5GEA 0 GBP
COBBHC-2045-5G COBBHC-2045-1G
2-Amino-4-(4-fluorophenyl)thiazole
2-Amino-4-(4-fluorophenyl)thiazole
Formula: C₉H₇FN₂S
MW: 194.2287 g/mol
MDL Number: MFCD01086598
CAS Number: 77815-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR