2-Amino-4-(4-bromophenyl)thiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2098-5G HC-2098-100G HC-2098-25G
COBBHC-2098-5GEA 0 GBP
COBBHC-2098-5G COBBHC-2098-100G COBBHC-2098-25G
2-Amino-4-(4-bromophenyl)thiazole
2-Amino-4-(4-bromophenyl)thiazole
Formula: C₉H₇BrN₂S
MDL Number: MFCD00051953
CAS Number: 2103-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR