4-Trifluoromethoxythioanisole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl(4-(trifluoromethoxy)phenyl)sulphane, 4-(Trifluoromethoxy)phenyl methyl sulphphide, 4-(Trifluoromethoxy)phenyl methyl sulphide

OT-0531-5G OT-0531-25G OT-0531-1G
COBBOT-0531-5GEA 0 GBP
COBBOT-0531-5G COBBOT-0531-25G COBBOT-0531-1G
4-Trifluoromethoxythioanisole
4-Trifluoromethoxythioanisole
Formula: C₈H₇F₃OS
MW: 208.20 g/mol
MDL Number: MFCD01320776
CAS Number: 2546-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR