2-Bromo-N-(2,3-dimethylphenyl)benzenesulphonamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6119-5G
HC-6119-1G
COBBHC-6119-5GEA
0
GBP
COBBHC-6119-5G
COBBHC-6119-1G
2-Bromo-N-(2,3-dimethylphenyl)benzenesulphonamide
2-Bromo-N-(2,3-dimethylphenyl)benzenesulphonamide
Formula:
C₁₄H₁₄BrNO₂S |
MDL Number:
MFCD05617811 CAS Number: 941294-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...